Single element standard for AA and MP-AES, Cadmium
Proveedor: Agilent
5190-8270
HEWL5190-8270EA 31 EUR
HEWL5190-8270
Single element standard for AA and MP-AES, Cadmium
Normativas Estándares catiónicos Estándares de cadmio
Agilent offers a comprehensive range of single and multi-element standards, ionisation buffers and environmental standards for atomic spectroscopy.
  • Gives you confidence in your AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.