CE capillaries, alignment interfaces and capillary cassette
Proveedor: Agilent
G1600-60002 G1600-60150 G1600-60210 G1600-60230 G1600-60310 G1600-60330 G7100-60002
HEWLG1600-60002EA 344 EUR
HEWLG1600-60002 HEWLG1600-60150 HEWLG1600-60210 HEWLG1600-60230 HEWLG1600-60310 HEWLG1600-60330 HEWLG7100-60002
CE capillaries, alignment interfaces and capillary cassette
Sistemas de electroforesis Electroforesis capilar
Alignment interfaces are an integral part of the Agilent diode-array detection (DAD) system. These interfaces contain optical slits which are precisely matched to the capillary inner diameter for optimised sensitivity and linear detection range.
In combination with the capillary cassette, alignment interfaces simplify capillary exchange, protect the fragile detection window and ensure exact alignment of the window in the detector. Quick-change cassette allows capillary exchange in less than one minute.
Cassette and interfaces accept all commercially available capillaries (~365 μm Ext.Ø)More Product Information