Capillary columns, HayeSep Q
Proveedor: Agilent
G3591-70005 G3591-70006 G3591-70007 G3591-70011 G3591-81033 G3591-81047 G3591-81121 G3591-82002 G3591-82004 G3591-82020 G3591-82023 G3591-82033 G3591-82121 G3591-82159 G3591-80002 G3591-80004 G3591-80019 G3591-80020 G3591-80023 G3591-80033 G3591-80047 G3591-80121 G3591-81002 G3591-81004 G3591-81019 G3591-81020 G3591-81023
HEWLG3591-70005EA 306 EUR
HEWLG3591-70005 HEWLG3591-70006 HEWLG3591-70007 HEWLG3591-70011 HEWLG3591-81033 HEWLG3591-81047 HEWLG3591-81121 HEWLG3591-82002 HEWLG3591-82004 HEWLG3591-82020 HEWLG3591-82023 HEWLG3591-82033 HEWLG3591-82121 HEWLG3591-82159 HEWLG3591-80002 HEWLG3591-80004 HEWLG3591-80019 HEWLG3591-80020 HEWLG3591-80023 HEWLG3591-80033 HEWLG3591-80047 HEWLG3591-80121 HEWLG3591-81002 HEWLG3591-81004 HEWLG3591-81019 HEWLG3591-81020 HEWLG3591-81023
Capillary columns, HayeSep Q
Columnas de cromatografía
Packed GC columns are designed and manufactured to offer excellent and reproducible performance for all sample types associated with packed column separations, most important in the hydrocarbon processing industry.
  • Highly efficient and rigorous packing technology assures column-to-column reproducibility and ultimate efficiency
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance