Avantor® Hichrom HI-WAX Plus, GC Columns
Proveedor: Avantor
HI14-18-018-10EA 518 EUR
HI14-18-018-10 HI14-18-018-20 HI14-18-018-40 HI14-18-018-50 HI14-18-018-60 HI14-18-020-10 HI14-18-020-20 HI14-18-020-40 HI14-18-020-50 HI14-18-020-60 HI14-18-040-10 HI14-18-040-20 HI14-18-040-40 HI14-18-040-50 HI14-18-040-60 HI14-20-010-10 HI14-20-010-20 HI14-20-010-40 HI14-20-010-50 HI14-20-010-60 HI14-20-020-10 HI14-20-020-20 HI14-20-020-40 HI14-20-020-50 HI14-20-020-60 HI14-20-040-10 HI14-20-040-20 HI14-20-040-40 HI14-20-040-50 HI14-20-040-60 HI14-20-100-10 HI14-20-100-20 HI14-20-100-40 HI14-20-100-50 HI14-20-100-60 HI14-25-010-15 HI14-25-010-30 HI14-25-010-50 HI14-25-010-60 HI14-25-025-10 HI14-25-025-105 HI14-25-025-15 HI14-25-025-30 HI14-25-025-50 HI14-25-025-60 HI14-25-050-10 HI14-25-050-105 HI14-25-050-15 HI14-25-050-30 HI14-25-050-50 HI14-25-050-60 HI14-25-100-10 HI14-25-100-105 HI14-25-100-15 HI14-25-100-30 HI14-25-100-50 HI14-25-100-60 HI14-32-010-10 HI14-32-010-105 HI14-32-010-15 HI14-32-010-30 HI14-32-010-50 HI14-32-010-60 HI14-32-025-10 HI14-32-025-105 HI14-32-025-15 HI14-32-025-30 HI14-32-025-50 HI14-32-025-60 HI14-32-050-10 HI14-32-050-105 HI14-32-050-15 HI14-32-050-30 HI14-32-050-50 HI14-32-050-60 HI14-32-100-10 HI14-32-100-105 HI14-32-100-15 HI14-32-100-30 HI14-32-100-50 HI14-32-100-60 HI14-32-150-10 HI14-32-150-105 HI14-32-150-15 HI14-32-150-30 HI14-32-150-50 HI14-32-150-60 HI14-32-300-10 HI14-32-300-105 HI14-32-300-15 HI14-32-300-30 HI14-32-300-50 HI14-32-300-60 HI14-53-010-10 HI14-53-010-15 HI14-53-010-30 HI14-53-025-15 HI14-53-025-30 HI14-53-025-50 HI14-53-025-60 HI14-53-050-10 HI14-53-050-15 HI14-53-050-30 HI14-53-050-50 HI14-53-050-60 HI14-53-100-10 HI14-53-100-15 HI14-53-100-30 HI14-53-100-50 HI14-53-100-60 HI14-53-100-75 HI14-53-150-10 HI14-53-150-15 HI14-53-150-30 HI14-53-150-50 HI14-53-150-60 HI14-53-150-75 HI14-53-200-10 HI14-53-200-15 HI14-53-200-30 HI14-53-200-50 HI14-53-200-60 HI14-53-200-75 HI14-53-250-10 HI14-53-250-15 HI14-53-250-30 HI14-53-250-50 HI14-53-250-60 HI14-53-250-75 HI14-05-005-2 HI14-05-005-2.5 HI14-05-005-5 HI14-10-010-10 HI14-10-010-15 HI14-10-010-5 HI14-10-020-10 HI14-10-020-15 HI14-10-020-5 HI14-10-040-10 HI14-10-040-15 HI14-10-040-5 HI14-15-015-10 HI14-15-015-15 HI14-15-015-20 HI14-15-015-30 HI14-15-015-5 HI14-25-025-15G5 HI14-25-025-30G5 HI14-25-025-60G5 HI14-53-100-30G5
Avantor® Hichrom HI-WAX Plus, GC Columns
Columnas de cromatografía
Max temperature limits may change depending on the film thickness.
Order Now
ESPECIFICACIONES
Phase name HI-WAX Plus
Environmentally Preferable
Phase composition Polyethylene Glycol (PEG)
Polarity Polar stationary phase
Crossbond Yes
Código USP G14, G15, G16, G20, G39