Avantor® Hichrom HI-XMLB, GC Columns
Proveedor: Avantor
HI20-18-018-20EA 1100 EUR
HI20-18-018-20 HI20-18-018-30 HI20-20-033-25 HI20-25-010-15 HI20-25-010-30 HI20-25-015-30 HI20-25-015-60 HI20-25-025-15 HI20-25-025-30 HI20-25-025-60 HI20-25-050-30 HI20-25-100-30 HI20-32-025-30 HI20-32-025-60 HI20-32-050-30 HI20-32-050-60 HI20-53-150-15 HI20-53-150-30 HIGCHI20-18-018-40
Avantor® Hichrom HI-XMLB, GC Columns
Columnas de cromatografía
Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
  • Phase composition: crossbonded low-polarity proprietary phase
  • Long lifetime
Max temperature limits may change depending on the film thickness.
Order Now
ESPECIFICACIONES
Phase name HI-XMLB
Environmentally Preferable
Phase composition Proprietary Phase developed for semi-volatiles environmental - low bleeding
Polarity Low-polarity stationary phase
Crossbond Yes