Avantor® Hichrom HI-35 MS, GC Columns
Proveedor: Avantor
HI21-25-010-30G5EA 810 EUR
HI21-25-010-30G5 HI21-25-025-15G5 HI21-25-025-30G10 HI21-25-025-30G5 HI21-25-025-60G10 HI21-25-025-60G5 HI21-25-050-15G10 HI21-25-050-30G10 HI21-25-050-30G5 HI21-32-025-30G5 HI21-32-100-30G5 HI21-53-100-30G5 HI21-18-018-10 HI21-18-018-20 HI21-18-018-40 HI21-18-018-50 HI21-18-018-60 HI21-18-020-10 HI21-18-020-20 HI21-18-020-40 HI21-18-020-50 HI21-18-020-60 HI21-18-036-20 HI21-18-036-40 HI21-18-036-60 HI21-18-040-10 HI21-18-040-20 HI21-18-040-40 HI21-18-040-50 HI21-18-040-60 HI21-20-010-10 HI21-20-010-20 HI21-20-010-40 HI21-20-010-50 HI21-20-010-60 HI21-20-020-10 HI21-20-020-20 HI21-20-020-40 HI21-20-020-50 HI21-20-020-60 HI21-20-033-12 HI21-20-033-25 HI21-20-033-30 HI21-20-033-50 HI21-20-033-60 HI21-20-040-10 HI21-20-040-20 HI21-20-040-40 HI21-20-040-50 HI21-20-040-60 HI21-20-100-10 HI21-20-100-20 HI21-20-100-40 HI21-20-100-50 HI21-20-100-60 HI21-25-010-15 HI21-25-010-30 HI21-25-010-50 HI21-25-010-60 HI21-25-025-10 HI21-25-025-105 HI21-25-025-15 HI21-25-025-30 HI21-25-025-50 HI21-25-025-60 HI21-25-050-10 HI21-25-050-105 HI21-25-050-15 HI21-25-050-30 HI21-25-050-50 HI21-25-050-60 HI21-25-100-10 HI21-25-100-105 HI21-25-100-15 HI21-25-100-30 HI21-25-100-50 HI21-25-100-60 HI21-32-010-10 HI21-32-010-105 HI21-32-010-15 HI21-32-010-30 HI21-32-010-50 HI21-32-010-60 HI21-32-025-10 HI21-32-025-105 HI21-32-025-15 HI21-32-025-30 HI21-32-025-50 HI21-32-025-60 HI21-32-050-10 HI21-32-050-105 HI21-32-050-15 HI21-32-050-30 HI21-32-050-50 HI21-32-050-60 HI21-32-100-10 HI21-32-100-105 HI21-32-100-15 HI21-32-100-30 HI21-32-100-50 HI21-32-100-60 HI21-32-150-10 HI21-32-150-105 HI21-32-150-15 HI21-32-150-30 HI21-32-150-50 HI21-32-150-60 HI21-53-010-10 HI21-53-010-15 HI21-53-010-30 HI21-53-025-15 HI21-53-025-30 HI21-53-025-50 HI21-53-025-60 HI21-53-050-10 HI21-53-050-15 HI21-53-050-30 HI21-53-050-50 HI21-53-050-60 HI21-53-100-10 HI21-53-100-15 HI21-53-100-30 HI21-53-100-50 HI21-53-100-60 HI21-53-100-75 HI21-53-150-10 HI21-53-150-15 HI21-53-150-30 HI21-53-150-50 HI21-53-150-60 HI21-53-150-75 HI21-53-300-30 HI21-05-005-2 HI21-05-005-2.5 HI21-05-005-5 HI21-10-010-10 HI21-10-010-15 HI21-10-010-5 HI21-10-020-10 HI21-10-020-15 HI21-10-020-5 HI21-10-040-10 HI21-10-040-15 HI21-10-040-5 HI21-15-015-10 HI21-15-015-15 HI21-15-015-20 HI21-15-015-30 HI21-15-015-5 HIGCHI21-25-020-30
Avantor® Hichrom HI-35 MS, GC Columns
Columnas de cromatografía
Proprietary low polarity and low bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and also an excellent choice as a confirmation column for semi-volatile environmental analysis.
  • Phase composition: Cross-bonded low polarity proprietary phase
  • Long lifetime
Max. temperature limits may change depending on the film thickness.