Avantor® Hichrom HI-VOC 1, GC Columns
Proveedor: Avantor
HI64-25-100-30EA 1090 EUR
HI64-25-100-30 HI64-25-100-60 HI64-25-150-30 HI64-25-150-60 HI64-25-200-30 HI64-25-200-60 HI64-25-300-30 HI64-25-300-60 HI64-32-100-105 HI64-32-100-30 HI64-32-100-60 HI64-32-100-75 HI64-32-150-105 HI64-32-150-30 HI64-32-150-60 HI64-32-150-75 HI64-32-200-105 HI64-32-200-30 HI64-32-200-60 HI64-32-200-75 HI64-32-300-105 HI64-32-300-30 HI64-32-300-60 HI64-32-300-75 HI64-32-500-105 HI64-32-500-30 HI64-32-500-60 HI64-32-500-75 HI64-53-100-30 HI64-53-100-60 HI64-53-100-75 HI64-53-150-30 HI64-53-150-60 HI64-53-150-75 HI64-53-200-30 HI64-53-200-60 HI64-53-200-75 HI64-53-300-30 HI64-53-300-60 HI64-53-300-75 HI64-53-500-30 HI64-53-500-60 HI64-53-500-75
Avantor® Hichrom HI-VOC 1, GC Columns
Columnas de cromatografía
Application specifc columns for volatile organic compounds (VOCs), solvents and purgeable compounds. Due to the high max. temperature, they are ideal for two-parallel columns confg. in the same oven. A good alternative to the HI-624 giving different selectivity. It is a more robust than the HI-624 and can operate at higher temperatures.
Max temperature limits may change depending on the film thickness.