Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Proveedor: Avantor
HI66-25-015-15EA 873 EUR
HI66-25-015-15 HI66-25-015-25 HI66-25-015-30 HI66-25-015-50 HI66-25-015-60 HI66-25-025-15 HI66-25-025-25 HI66-25-025-30 HI66-25-025-50 HI66-25-025-60 HI78-10-010-10 HI78-10-010-5
Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Columnas de cromatografía
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.