PEEKsil™, Chromatography Tubing, Precut
Proveedor: Avantor
HICH05-SS-05EA 40.3 EUR
HICH05-SS-05 HICH05-SS-10 HICH10-SS-07 HICH10-SS-10 HICH10-SS-30 HICH20-SS-10
PEEKsil™, Chromatography Tubing, Precut
Tuberías Chromatography Tubing
316 Stainless steel, precision bore ¹/₁₆" type tubing.
  • Passivated for chemical resistance
  • Square, burr-free ends
  • Precut lengths
0,005 and 0,007" Int.Ø- suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
0,010" Int.Ø - suitable for connection of analytical and semi-preparative columns containing 5 µm and 10 µm material. It should be used in all parts of the system where dead volume needs to be kept to minimum.