Tryptone Peptone Glucose Yeast Extract Broth Base
Proveedor: HIMEDIA
M969-500G
HIMEM969-500GEA 115 EUR
HIMEM969-500G
Tryptone Peptone Glucose Yeast Extract Broth Base
Medios Medios deshidratados para microbiología
This broth base without trypsin is used to test toxicity type of Clostridium botulinum cultures.