Measuring cylinders, class A, borosilicate glass, with single certificate
Proveedor: HIRSCHMANN
224015408 224016008 224017008 224017508 224018008 224018608 224019008 224019308 224019408
HIRS224015408EA 41.1 EUR
HIRS224015408 HIRS224016008 HIRS224017008 HIRS224017508 HIRS224018008 HIRS224018608 HIRS224019008 HIRS224019308 HIRS224019408
Measuring cylinders, class A, borosilicate glass, with single certificate
Probetas Probetas graduadas
DURAN®, borosilicate glass, tall form, class A.
  • Calibrated to contain 'ln', certificate of conformity (DE-M marking) with dated batch identification via website: internet: (www.hirschmannlab.de)
  • Hexagonal glass base, with spout
  • Blue graduations with ring marks at major graduations
With single certificate.