Measuring cylinders, tall form, class A, borosilicate glass, with DKD certificate
Proveedor: HIRSCHMANN
2240175DKD 2240180DKD 2240190DKD 2240193DKD 2240154DKD 2240160DKD 2240170DKD 2240194DKD
HIRS2240175DKDEA 94.3 EUR
HIRS2240175DKD HIRS2240180DKD HIRS2240190DKD HIRS2240193DKD HIRS2240154DKD HIRS2240160DKD HIRS2240170DKD HIRS2240194DKD
Measuring cylinders, tall form, class A, borosilicate glass, with DKD certificate
Probetas Probetas graduadas
DURAN® Borosilicate glass, Class A.
  • Blue graduation, main point ring graduation
  • Hexagonal glass base, spout
  • With dated batch identification
With DAkkS-Certificate.