Measuring cylinders, tall form, class A, PMP
Proveedor: HIRSCHMANN
7520170 7520175 7520180 7520186 7520190 7520193 7520194
HIRS7520170EA 24.7 EUR
HIRS7520170 HIRS7520175 HIRS7520180 HIRS7520186 HIRS7520190 HIRS7520193 HIRS7520194
Measuring cylinders, tall form, class A, PMP
Probetas Probetas graduadas
PMP, transparent, tall form, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • With embossed scale
  • Autoclavable - Does not affect accuracy
Certificaciones: DIN 12 681 / ISO 6706