2-Propanol, SemiConductor Grade (Honeywell 17829)

Proveedor: Honeywell Chemicals
Peligro

Sinónimos: Alcohol isopropílico, IPA

40219H-5L
HONC40219H-5LEA 312 EUR
HONC40219H-5L
2-Propanol, SemiConductor Grade (Honeywell 17829)
2-Propanol

Beilstein Number: 635639

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Pto. Ebullición: 77…83 °C (1.013 hPa)
Pto. de fusión: –90 °C
Densidad: 0.784…0.786 g/cm³ (20 °C)
Pto. de inflamación: 12 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011674
Núm. CAS: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
Índice Merck: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Assay (GC) Min. 99.8 %
Boiling range 81 - 83 °C
Density (D 20/4) 0.785 - 0.787
Refractive index (n 20/D) 1.3770 - 1.3780
Non-volatile matter Max. 5 ppm
Water (Karl Fischer) Max. 500 ppm
Free acid (as C₂H₅COOH) Max. 10 ppm
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.01 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Gallium (Ga) Max. 0.02 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.2 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.02 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.02 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 1 ppm
Sulfate (SO₄) Max. 1 ppm
Acetone (GC) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR