Ácido sulfúrico 95-97%, Semiconductor grade (Honeywell 17831)

Proveedor: Honeywell Chemicals
Peligro

40254H-2.5L
HONC40254H-2.5LEA 141 EUR
HONC40254H-2.5L
Ácido sulfúrico 95-97%, Semiconductor grade (Honeywell 17831)
Ácido sulfúrico

Beilstein Number: 2037554

Formula: H₂SO₄
Pto. Ebullición: 310 °C (1,013 hPa)
Pto. de fusión: –14…–10 °C
Densidad: 1,840 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064589
Núm. CAS: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Resultados test especificaciones

Assay 95 - 97 %
Residue on ignition Max. 5 ppm
Ammonium (NH₄) Max. 2 ppm
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Gallium (Ga) Max. 0.02 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.2 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.02 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.02 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 0.1 ppm
Nitrate (NO₃) Max. 0.2 ppm
Phosphate (PO₄) Max. 0.5 ppm
KMnO₄ reducing matter (as O) Max. 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR