Alternate Metals I for AA and ICP-OES
Proveedor: Honeywell Chemicals
E2169-100ML
HONCE2169-100MLEA 689 EUR
HONCE2169-100ML
Alternate Metals I for AA and ICP-OES
Normativas Estándares catiónicos Estándares de multielementos
Certified Reference Material (CRM).
20 mg/L each of Al, Fe, V, 10 mg/L each of Co, Cu, Mn, Ni, Zn, 5 mg/L each of Be, Sb, Tl in HNO₃ 2%