Analytical standard, Tetracyclin Hydrochlorid, reference material, Fluka™

Proveedor: Honeywell Chemicals

R1773-100MG
HONCR1773-100MGEA 40.8 EUR
HONCR1773-100MG
Analytical standard, Tetracyclin Hydrochlorid, reference material, Fluka™
Normativas Estándares orgánicos

Order Now

ESPECIFICACIONES

Descripción Tetracyclin Hydrochlorid
Environmentally Preferable
CAS No. 64-75-5
Formula C₂₂H₂₄N₂O₈ · HCl
Temperatura de almacenamiento 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR