Circulating cooling thermostats with natural refrigerant, Minichiller and Unichillers, with OLÉ controller
Proveedor: HUBER
3006.0091.98 3006.0089.98 3006.0090.98 3006.0092.98 3009.0090.98 3010.0050.98 3012.0120.98 3012.0126.98 3012.0142.98 3010.0103.98 3012.0260.98 3052.0012.98 3009.0244.98 3052.0018.98 3051.0020.98 3065.0001.98 3066.0002.98 3066.0003.98 3066.0004.98 3067.0001.98 3009.0200.98 3052.0020.98 3050.0003.98 3052.0024.98 3010.0130.98 3010.0124.98 3051.0018.98 3050.0012.98 3050.0026.98 3051.0003.98 3066.0001.98 3012.0263.98 3079.0006.98
HUBE3006.0091.98EA 5160 EUR
HUBE3006.0091.98 HUBE3006.0089.98 HUBE3006.0090.98 HUBE3006.0092.98 HUBE3009.0090.98 HUBE3010.0050.98 HUBE3012.0120.98 HUBE3012.0126.98 HUBE3012.0142.98 HUBE3010.0103.98 HUBE3012.0260.98 HUBE3052.0012.98 HUBE3009.0244.98 HUBE3052.0018.98 HUBE3051.0020.98 HUBE3065.0001.98 HUBE3066.0002.98 HUBE3066.0003.98 HUBE3066.0004.98 HUBE3067.0001.98 HUBE3009.0200.98 HUBE3052.0020.98 HUBE3050.0003.98 HUBE3052.0024.98 HUBE3010.0130.98 HUBE3010.0124.98 HUBE3051.0018.98 HUBE3050.0012.98 HUBE3050.0026.98 HUBE3051.0003.98 HUBE3066.0001.98 HUBE3012.0263.98 HUBE3079.0006.98
Circulating cooling thermostats with natural refrigerant, Minichiller and Unichillers, with OLÉ controller
Termostatos con circulación Sistemas de circulación cerrada Circuladores de enfriamiento
Many applications require a reliable source of cooling. Circulating chillers in the Unichiller® range offer an ideal solution for environmentally friendly and economical cooling in laboratory and industry. There are over 50 air and water cooled models to choose from, with cooling powers from 0,3 to 50 kW. Efficient energy management ensures low operating costs and reduced use of valuable fresh water. These circulating chillers are a resource saving solution which give a quick return on investment.
  • Robust stainless steel construction, ideal for simple applications
  • Large OLED display