Tubing, insulated/drain tubing, pressureless
Proveedor: HUBER
6084 6085 6427 6136 9277 6070 6071 6072 6073 6075 6076 6077 6079 6255 6350 6351 6386 6426 6428 6488 6655 6656 6657 6658 6784 6785 6786 6787 9274 9275 9276 9325 9326 9327 9328 9608 9609 9611 34322 9613 9610 9614 9612 6435 9854
HUBE6084EA 430 EUR
HUBE6084 HUBE6085 HUBE6427 HUBE6136 HUBE9277 HUBE6070 HUBE6071 HUBE6072 HUBE6073 HUBE6075 HUBE6076 HUBE6077 HUBE6079 HUBE6255 HUBE6350 HUBE6351 HUBE6386 HUBE6426 HUBE6428 HUBE6488 HUBE6655 HUBE6656 HUBE6657 HUBE6658 HUBE6784 HUBE6785 HUBE6786 HUBE6787 HUBE9274 HUBE9275 HUBE9276 HUBE9325 HUBE9326 HUBE9327 HUBE9328 HUBE9608 HUBE9609 HUBE9611 HUBE34322 HUBE9613 HUBE9610 HUBE9614 HUBE9612 HUBE6435 HUBE9854
Tubing, insulated/drain tubing, pressureless
Tuberías Tubo de transferencia
Insulated smooth silicon hose, ends covered with silicon caps, fixed length and drain tubing with variable length.