Upright Shelves, Norm 5
Proveedor: HUPFER METALLWERKE
0101325 0101295 0101315
HUPF0101325EA 168 EUR
HUPF0101325 HUPF0101295 HUPF0101315
Upright Shelves, Norm 5
Mobiliario Estantes
Modular shelving system made of high-quality stainless steel 1,4301 in hygienic design as per DIN 18868-2, suitable for continuous use at –40 to +60 °C (–40 to +140 °F), for arrangement in a straight line or for angled setup with corner hooks, freely extendable lengthwise.
  • Stainless steel
  • Pitch 150 mm
Uprights made of 25×25 mm square tubes, connected and stabilised by press-fitted 50×2 mm upright spokes running crosswise. Upper covering caps made of plastic, height-adjustable screw feet for compensating floor unevennesses of up to 25 mm as hygienicMore Product Information