Trypsin Protease, HyClone™
Proveedor: HyClone products (Cytiva)
SH30236.01 SH30236.02 SH30042.01 SH30042.02 SV30031.01 SV30037.01
HYCLSH30236.01EA 13.8 EUR
HYCLSH30236.01 HYCLSH30236.02 SH30042.01 HYCLSH30042.02 HYCLSV30031.01 HYCLSV30037.01
Trypsin Protease, HyClone™
Suplementos y aditivos de cultivo celular
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
Order Now
ESPECIFICACIONES
Environmentally Preferable
Especies
Fuente