Sodio piruvato 0.1 mol/l aqueous solution

Proveedor: HyClone products (Cytiva)

Sinónimos: Piruvato de sodio

SH30239.01
HYCLSH30239.01EA 19 EUR
HYCLSH30239.01
Sodio piruvato 0.1 mol/l aqueous solution
Sodio piruvato

Sodium pyruvate solution is an easily accessible carbohydrate source that may also have protective effects against hydrogen peroxide.

Formula: CH₃COCOONa
MW: 110.04 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00002586
Núm. CAS: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR