GC liners, LinerGOLD™
Proveedor: Thermo Fisher Scientific
453A2265-UI 453A1262-UI 453A1295-UI 453A2292-UI 453A1345-UI 453A2342-UI 453A1925-UI 453A2922-UI 453A1355-UI 453A2352-UI 453A1255-UI 453A1252-UI 453A1335-UI 453A2332-UI 45350030-UI 45350032-UI 45350033-UI 453T1905-UI 453T2999-UI 453T2845-UI
HYPE453A2265-UIEA 119 EUR
HYPE453A2265-UI HYPE453A1262-UI HYPE453A1295-UI HYPE453A2292-UI HYPE453A1345-UI HYPE453A2342-UI HYPE453A1925-UI HYPE453A2922-UI HYPE453A1355-UI HYPE453A2352-UI HYPE453A1255-UI HYPE453A1252-UI HYPE453A1335-UI HYPE453A2332-UI HYPE45350030-UI HYPE45350032-UI HYPE45350033-UI HYPE453T1905-UI HYPE453T2999-UI HYPE453T2845-UI
GC liners, LinerGOLD™
Suministros de cromatografía Inlet Liners de cromatografía
LinerGOLD™ GC liners provide exceptional inertness, leading to increased accuracy and precision by preventing loss of sensitive compounds.
  • Lower detection limits for a wide range of active compounds