SPE plates, 96-well, HyperSep™, Silica
Proveedor: Thermo Fisher Scientific
60300-482 60300-485 60300-486 60300-488
HYPE60300-482EA 399 EUR
HYPE60300-482 HYPE60300-485 HYPE60300-486 HYPE60300-488
SPE plates, 96-well, HyperSep™, Silica
Microplacas Placas de extracción en fase sólida
Thermo Scientific™ HyperSep Silica SPE 96-Well Plates and Individual Wells feature a polar sorbent primarily used to retain analytes in non polar matrices, and can effectively separate compounds of very similar structure.
  • Extraction of polar compounds, carotenoids, fatty acids