Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Proveedor: Thermo Fisher Scientific
6ACV20-1PE 6ACV20-1PEF 6ACV20-1PET 6ACV20-1R 6ACV20-1RF 6ACV20-1RT 6ACV20-2PE 6ACV20-2PET
HYPE6ACV20-1PEEA 53.8 EUR
HYPE6ACV20-1PE HYPE6ACV20-1PEF HYPE6ACV20-1PET HYPE6ACV20-1R HYPE6ACV20-1RF HYPE6ACV20-1RT HYPE6ACV20-2PE HYPE6ACV20-2PET
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Viales
Use Thermo Scientific™ SureSTART™ 20 ml glass crimp top headspace vials in your high-throughput volatile gas analyses. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC and single or triple quadrupole MS systems.
These headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyses. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure theirMore Product Information