Glass screw top vials, 4 ml, level 2 high-throughput applications, SureSTART™
Proveedor: Thermo Fisher Scientific
6ASV13-1P 6ASV13-2P
HYPE6ASV13-1PEA 29 EUR
HYPE6ASV13-1P HYPE6ASV13-2P
Glass screw top vials, 4 ml, level 2 high-throughput applications, SureSTART™
Viales
Use Thermo Scientific™ SureSTART™ 4 ml glass screw top headspace vials, performance level 2, in your high-throughput volatile gas analyses. Our screw top headspace vials are a novelty in a crimp-neck dominated field.
They provide a ready-to-use, convenient solution that does not require crimping or decrimping tools. These are performance level 2 vials that ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems. The headspace vials are made to withstand higher temperatures and internalMore Product Information