ADP-Na3 ≥95%, en polvo

Proveedor: MP Biomedicals

Sinónimos: Adenosine-5'-Diphosphate Trisodium Salt

02100049.5 0210004910 0210004901 0210004905
ICNA02100049.5EA 66.7 EUR
ICNA02100049.5 ICNA0210004910 ICNA0210004901 ICNA0210004905
ADP-Na3 ≥95%, en polvo
ADP-Na3

Soluble in water (50 mg/ml); practically insoluble in methanol, ethanol, and ether.


Adenosine 5′-diphosphate (ADP) is an adenine nucleotide involved in energy storage and nucleic acid metabolism via its conversion into ATP by ATP synthases. ADP affects platelet activation through its interaction with ADP receptors P2Y1, P2Y12 and P2X1. Upon its conversion to adenosine by ecto-ADPases, platelet activation is inhibited via adenosine receptors.

Formula: C₁₀H₁₂N₅Na₃O₁₀P₂
Núm. CAS: 2092-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR