1-Naphthyl phosphate disodium salt monohydrate, crystalline

Proveedor: MP Biomedicals
Atención

02100257.1 02100257.2 0210025701 0210025705
ICNA02100257.1EA 33.9 EUR
ICNA02100257.1 ICNA02100257.2 ICNA0210025701 ICNA0210025705
1-Naphthyl phosphate disodium salt monohydrate, crystalline
1-Naphthyl phosphate disodium salt monohydrate

α - Naphthyl phosphate disodium salt is a substrate for determination of phosphatase.


1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.


Soluble in water (50 mg/ml)

Formula: C₁₀H₇Na₂O₄P·1H₂O
Pto. de fusión: 157…159 °C (lit.)
Temperatura de almacenaje: Congelador
Número MDL: MFCD00150614
Núm. CAS: 207569-06-0
EINECS: 218-564-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR