(±)-Ácido 2-aminobutírico, en polvo

Proveedor: MP Biomedicals

0210043090 0210043025 0210043080
ICNA0210043090EA 847 EUR
ICNA0210043090 ICNA0210043025 ICNA0210043080
(±)-Ácido 2-aminobutírico, en polvo
(±)-Ácido 2-aminobutírico

Soluble in water (5%). Sparingly soluble in alcohol; insoluble in ether.

Formula: C₄H₉NO₂
Número MDL: MFCD00008093
Núm. CAS: 2835-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR