Coenzyme A trilithium salt dihydrate 96, powder_crystalline_white

Proveedor: MP Biomedicals

Sinónimos: Ubidecarenone (coenzyme, q-10), CoA Li3, Trilithium coenzyme A

0210049390 0210049310 0210049325 0210049350 0210049380 0210049383 0210049391
ICNA0210049390EA 1308 EUR
ICNA0210049390 ICNA0210049310 ICNA0210049325 ICNA0210049350 ICNA0210049380 ICNA0210049383 ICNA0210049391
Coenzyme A trilithium salt dihydrate 96, powder_crystalline_white
Coenzyme A trilithium salt dihydrate

Coenzyme A is an essential cofactor in enzymatic acetyl transfer reactions. The principal biologically active forms of pantothenic acid are coenzyme A (CoA) and acyl carrier protein (ACP). In CoA, the business center of the molecule is the pantothenic acid metabolite 4'-phosphopantetheine. Coenzyme A is comprised of 4'-phosphopantetheine linked by an anhydride bond to the nucloetide adenosine 5'-monophosphate. 4'-Phosphopantetheine itself is comprised of pantothenic acid linked at one end, via an amide bond, to beta-mercaptoethylamine, derived from L-cysteine, and at the other end to a phosphate group. The sulfhydryl group of 4'-phosphopantetheine, which is the business end of the coenzyme, forms thioesters with acyl groups producing acyl-CoA derivatives, including acetyl-CoA.
Coenzyme A facilitates removal of lipid peroxides by increasing mobilization of fatty acids, and promote repair of plasma membranes by activating phospholipid synthesis.


  • Presentation: White Crystalline Powder
  • Soluble in water (50 mg/mL - clear, colorless to faint yellow solution)
  • Storage temperature: -20 °C

Formula: C₂₁H₃₃N₇O₁₆P₃SLi₃·2H₂O
MW: 821.4 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00075848
Núm. CAS: 18439-24-2

Order Now

Resultados test especificaciones

Identity Test Passes
Purity ≥96%
OD250/OD260 (pH 2) 0.80-0.89


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR