Estradiol ≥98%, white powder

Proveedor: MP Biomedicals
Peligro

Sinónimos: beta-Estradiol, 17β-Estradiol

02101656.2 0210165601 0210165605 0210165625
ICNA02101656.2EA 86.1 EUR
ICNA02101656.2 ICNA0210165601 ICNA0210165605 ICNA0210165625
Estradiol ≥98%, white powder
Estradiol

β-Estradiol is used to study cell differentiation and transformations (tumorigenicity). The pharmacology and HPLC determination of β-Estradiol in plasma and urine have been reported.


β-Estradiol is used to study cell differentiation and transformations (tumorigenicity). The pharmacology and HPLC determination of β-Estradiol in plasma and urine have been reported.


β-Estradiol is almost insoluble in water. Freely soluble in alcohol; soluble in acetone, dioxane, other organic solvents; solutions of fixed alkali hydroxides; sparingly soluble in vegetable oils; soluble in methanol : CHCl3 (1:1). Dissolve 50 mg/ml in ethanol with heating and observed a clear, colourless solution. It is also soluble in acetone and dioxane but only sparingly soluble in vegetable oils.


Precaución: Research Use Only (RUO).

Formula: C₁₈H₂₄O₂
MW: 272.39 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003693
Núm. CAS: 50-28-2
EINECS: 200-023-8

Order Now

Resultados test especificaciones

Identity Test Passes
Assay ≥98%
FTIR Conforms to Standard
Melting Point 173 - 179 °C
Optical Rotation +76° to +83° (c = 1% in dioxane at 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR