Sulphabenzamide, powder

Proveedor: MP Biomedicals

Sinónimos: 4-Amino-N-benzoylbenzenesulphonamide, N-(4-Aminobenzenesulphonyl)benzamide, N-Benzoylsulphanilamide

0210298125
ICNA0210298125EA 55.3 EUR
ICNA0210298125
Sulphabenzamide, powder
Sulphabenzamide
Formula: C₁₃H₁₂N₂O₃S
MW: 276.31 g/mol
Número MDL: MFCD00044890
Núm. CAS: 127-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR