DL-Treonina ≥99%, white powder

Proveedor: MP Biomedicals

0210305010 0210305080 0210305025 0210305091
ICNA0210305010EA 39.4 EUR
ICNA0210305010 ICNA0210305080 ICNA0210305025 ICNA0210305091
DL-Treonina ≥99%, white powder
DL-Treonina

DL-Threonine is an essential α - amino acid classified as polar which has two chiral centers. In plants and micro-organisms, threonine is synthesized from aspartic acid via aspartyl - semialdehyde and homoserine.


DL-Threonine is as an additive to animal feed. They are also used to chelate metal cations in order to improve the absorption of minerals from supplements, which may be required to improve the health or production of animals. This chelating ability is also used in fertilizers for agriculture.


DL-Threonine is as an additive to animal feed. They are also used to chelate metal cations in order to improve the absorption of minerals from supplements, which may be required to improve the health or production of animals. This chelating ability is also used in fertilizers for agriculture.


Soluble in water (50 mg/mL- clear, colorless solution).

Formula: C₄H₉NO₃
Pto. de fusión: 238…248 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00063722
Núm. CAS: 80-68-2
EINECS: 201-300-6

Order Now

Resultados test especificaciones

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Loss on Drying ≤0.2%
Optical Rotation -1° to +1° (c=3, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR