Ácido 8-anilinonaftaleno-1-sulfónico, dark powder

Proveedor: MP Biomedicals
Atención

0210563301 0210563305 0210563310 0210563325
ICNA0210563301EA 37 EUR
ICNA0210563301 ICNA0210563305 ICNA0210563310 ICNA0210563325
Ácido 8-anilinonaftaleno-1-sulfónico, dark powder
Ácido 8-anilinonaftaleno-1-sulfónico

8-Anilino-1-naphthalene Sulphonic Acid is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins.


ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants. It is a hydrophobic fluorescent probe for undenatured proteins on polyacrylamide gels.


Soluble in DMF (50 mg/ml). Soluble in 1 N NaOH, methanol (50 mg/ml - clear brownish green solution), chloroform (30 mg/ml in a 1:1 mixture produces a clear olive green solution); slightly water soluble.


Precaución: Research Use Only (RUO).

Formula: C₁₆H₁₃NO₃S
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003998
Núm. CAS: 82-76-8

Order Now

Resultados test especificaciones

Identity Test Passes
FTIR Conforms to Standard

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR