Ácido cítrico monohidrato, white crystalline powder

Proveedor: MP Biomedicals
Atención

02150699.5 0215069901 0215069905
ICNA02150699.5EA 45.6 EUR
ICNA02150699.5 ICNA0215069901 ICNA0215069905
Ácido cítrico monohidrato, white crystalline powder
Ácido cítrico monohidrato

Citric acid acts as an iron chelator. It can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications. In animals, citric acid improves the utilization of nutritional calcium. It is a useful buffer component.


Soluble in water, methanol.

Formula: C₆H₈O₇.H₂O
Número MDL: MFCD00149972
Núm. CAS: 5949-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR