α-Ketoglutaric acid disodium salt dihydrate, white crystalline powder

Proveedor: MP Biomedicals

Sinónimos: 2-Oxoglutaric acid disodium salt dihydrate, α-Ketoglutaric acid disodium salt dihydrate, Sodium α-ketoglutarate dibasic dihydrate

0215139383 0215139390 0215139350
ICNA0215139383EA 479 EUR
ICNA0215139383 ICNA0215139390 ICNA0215139350
α-Ketoglutaric acid disodium salt dihydrate, white crystalline powder
α-Ketoglutaric acid disodium salt dihydrate

α-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) is the hydrated disodium salt of 2-oxoglutaric acid.


α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry. α-Ketoglutaric acid disodium salt dihydrate (2-Oxoglutaric acid disodium salt) may be used as starting reagent for the synthesis of deuteriated 2-oxo[3,3-2H2]glutarate disodium salt.


Soluble in water (1,0 M - clear, colourless solution).

Formula: C₅H₄O₅·2Na·2H₂O
Temperatura de almacenaje: Nevera
Número MDL: MFCD00150702
Núm. CAS: 305-72-6
EINECS: 206-167-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR