Nalidixic acid sodium salt

Proveedor: MP Biomedicals

0215172401 0215172405 0215172425 0215172480
ICNA0215172401EA 27.9 EUR
ICNA0215172401 ICNA0215172405 ICNA0215172425 ICNA0215172480
Nalidixic acid sodium salt
Nalidixic acid sodium salt

Nalidixic acid sodium salt is an antibacterial agent (synthetic quinolone antibiotic), which is inhibitor of bacterial DNA polymerase (DNA gyrase) & avian myeloblastoma virus reverse transcriptase.


Nalidixic acid sodium salt is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. Inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.


Typically used at a concentration of 15 ug/ml in molecular biology.


Soluble in water (100 mg/ml - clear, light yellow solution). Stock solutions can be aliquoted and stored at -20 °C for up to approximately 3 to 4 months.

Formula: C₁₂H₁₁N₂NaO₃
Número MDL: MFCD00064376
Núm. CAS: 3374-05-8

Order Now

Resultados test especificaciones

Form Off-white powder
Storage Store at 0 °C, Use only in area provided with appropriate exhaust ventilation
Moisture Content ≤9%

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR