Safranina O, dark green powder C.I. 50240

Proveedor: MP Biomedicals
Atención

0215204005 0215204025 0215204080
ICNA0215204005EA 38.3 EUR
ICNA0215204005 ICNA0215204025 ICNA0215204080
Safranina O, dark green powder C.I. 50240
Safranina O

Safranine O is a redox and adsorption indicator.


Safranine O is a biological stain used in histology, cytology, bacteriology.


Soluble in ethanol 50% (Clear, orange-red solution with fluorescence).

Formula: C₂₀H₁₉ClN₄
Densidad: 1,447 g/cm³ (22 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011759
Núm. CAS: 477-73-6
EINECS: 207-518-8

Order Now

Resultados test especificaciones

Identity Test Passes
UV/Vis: l max (50% ethanol) 530 ± 5 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR