Amonio hierro (II) sulfato hexahidrato 98.5-101.5%, crystalline ACS

Proveedor: MP Biomedicals
Atención

0215252390 0215252391
ICNA0215252390EA 81.5 EUR
ICNA0215252390 ICNA0215252391
Amonio hierro (II) sulfato hexahidrato 98.5-101.5%, crystalline ACS
Amonio hierro (II) sulfato hexahidrato

Ferrous ammonium sulphate is often employed as an analytical standard, and has been used in a variety of other applications from nanomaterials to general redox reactions. It is used for titration purposes.


Soluble in water; practically insoluble in alcohol.


Precaución: Research Use Only (RUO).

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00150530
Núm. CAS: 7783-85-9

Order Now

Resultados test especificaciones

Identity Test Passes
Assay 98,5 - 101,5%
Insoluble Matter ≤0,01%
Phosphate (PO4) <0,003%
Copper (Cu) <0,003%
Manganese (Mn) <0,01%
Zinc (Zn) 0,003%
Ferric Ion (Fe3+) 0,01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR