Human beta2-Microglobulin (from Urine), MP Biomedicals

Proveedor: MP Biomedicals

02153903.5
ICNA02153903.5EA 505 EUR
ICNA02153903.5
Human beta2-Microglobulin (from Urine), MP Biomedicals
Proteínas y Péptidos

  • Lyophilized
  • Purity: 30 to 60% total protein


Storage temperature: -20 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR