Ácido algínico, M.W. ~240,000 (de Macrocystis pyrifera (kelp)), off-white powder

Proveedor: MP Biomedicals

0215472291 0215472280 0215472283 0215472290
ICNA0215472291EA 0 EUR
ICNA0215472291 ICNA0215472280 ICNA0215472283 ICNA0215472290
Ácido algínico, M.W. ~240,000 (de Macrocystis pyrifera (kelp)), off-white powder
Ácido algínico

A straight-chain, hydrophilic, colloidal, polyuronic acid composed primarily of anhydro-β-D-mannuronic acid residues with 1→4 linkage.


Alginic acid is suitable reagent used to study the in-vitro and in-vivo mitogenic activity of alginates. It is suitable for use in the spectrophotometric determination of transparent exopolymer particles by dye-binding assay.


It is reported to inhibit the reverse transcriptase activity of HIV. Structural investigation of alginic acid, a marine algal polysaccharide poly-α-L-guluronic acid, has been reported. Anti-inflammatory effect of alginic acid, isolated from the brown algae Sargassum wightii, in type II collagen induced arthritic rats has been reported. Pyrolysis of alginic acid using py-GC/MS over a wide temperature range (200-800°C) and thermogravimetric analysis (TGA) has been reported. It was reported to cause the biocorrosion of thin films of copper on germanium internal reflection elements (IREs).


Insoluble, but swells in water; slightly soluble in alkalis [soluble in 0,1 M NaOH].

Formula: (C₆H₈O₆)n
Pto. de fusión: 300 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00081309
Núm. CAS: 9005-32-7
EINECS: 232-680-1

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR