Nisin (de Lactococcus lacti)

Proveedor: MP Biomedicals

0215583905 0215583925
ICNA0215583905EA 142 EUR
ICNA0215583905 ICNA0215583925
Nisin (de Lactococcus lacti)
Nisin

Nisin is a polycyclic lantibiotic produced by Lactococcus lactis. It is an antibiotic with bactericidal action and is commonly used as a food preservative. It is also used as a selective agent in cell culture for the isolation of gram-negative bacteria, yeast, and moulds.


Mode of action: Binds to and interferes with the permeability of the cytoplasmic membrane.
Antimicrobial spectrum: Gram-negative and gram-positive bacteria (requires chelator such as EDTA.


Soluble in dilute acids such as 0,02 N HCl (118 mg/ml). Any undissolved material is the dairy solids and should not be considered a problem. Stable to boiling in acid solutions.

Formula: C₁₄₃H₂₃₀N₄₂O₃₇S₇
Número MDL: MFCD00131724
Núm. CAS: 1414-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR