Primaquine diphosphate, dark orange powder

Proveedor: MP Biomedicals
Peligro

Sinónimos: N4-(6-Methoxyquinolin-8-yl)pentane-1,4-diamine bis(phosphate), NSC 149765, Primaquine bisphosphate

0215636605 0215636625 0215636650 0215636610
ICNA0215636605EA 151 EUR
ICNA0215636605 ICNA0215636625 ICNA0215636650 ICNA0215636610
Primaquine diphosphate, dark orange powder
Primaquine diphosphate

Soluble in water (50 mg/ml).


Soluble in water (50 mg/ml).

Formula: C₁₅H₂₇N₃O₉P₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00013166
Núm. CAS: 63-45-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR