di-Sodio tetraborato decahidrato ~99%, powder_white

Proveedor: MP Biomedicals
Peligro

Sinónimos: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

02190308.5
ICNA02190308.5EA 38.1 EUR
ICNA02190308.5
di-Sodio tetraborato decahidrato ~99%, powder_white
di-Sodio tetraborato decahidrato

Borax is a salt of boric acid which is used as a buffer. It has a co-complexing ability with other agents in water to form complex ions with various substances. It is also commonly utilized in chromatography and electrophoresis studies.


Borax is typically used as a buffer component, manufacturing of glazes and enamels, fireproofing fabrics and wood, antiseptic and detergents.


Precaución: Research Use Only (RUO).

Formula: Na₂B₄O₇·10H₂O
MW: 381.37 g/mol
Pto. Ebullición: 1575 °C (lit.)
Pto. de fusión: 741 °C
Densidad: 1.730 g/mL (lit.)
Pto. de inflamación: 1575 °C (lit.)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00149193
Núm. CAS: 1303-96-4
EINECS: 215-540-4
Índice Merck: 13,08662

Order Now

Resultados test especificaciones

Identity Test Passes
Purity ~99%
Moisture Content 30-40%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR