Cloruro de 3-metilamino-7-dimetilaminofenotiazin-5-io, dark green powder, certificado C.I. 52010

Proveedor: MP Biomedicals

0219052001 0219052010
ICNA0219052001EA 37.1 EUR
ICNA0219052001 ICNA0219052010
Cloruro de 3-metilamino-7-dimetilaminofenotiazin-5-io, dark green powder, certificado C.I. 52010
Cloruro de 3-metilamino-7-dimetilaminofenotiazin-5-io

Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.


Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.


Soluble in water (50 mg/ml) or ethanol (8-10 mg/ml); insoluble in diethyl ether.

Formula: C₁₅H₁₆ClN₃S
Pto. de fusión: 205…210 °C (desc., lit.)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011935
Núm. CAS: 531-55-5
EINECS: 208-511-2

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR