Aceite de maíz (from corn), yellow oil
Proveedor: MP Biomedicals
0290141401 0290141405 0290141425
ICNA0290141401EA 47 EUR
ICNA0290141401 ICNA0290141405 ICNA0290141425
Aceite de maíz (from corn), yellow oil
Aceite de maíz
Corn oil is used as a research animal diet supplement. It is also used in research applications as delivery vehicle for fat soluble compounds including toxins.
Oleic acid: 43 - 47%, linoleic acid: 39 - 42%; palmitic acid: 7 - 8%; stearic acid: 3 - 4%; arachidic acid: 0,5%; approx. 7,6 lbs/gal (Lit.).
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00130865
Núm. CAS: 8001-30-7
Order Now
Resultados test especificaciones
Identity Test Passes