Sodio nitrito 97.0-101.0%, pale yellow powder USP

Proveedor: MP Biomedicals
Peligro

Sinónimos: Diazotizing salts

0296030190 0296030191
ICNA0296030190EA 70.7 EUR
ICNA0296030190 ICNA0296030191
Sodio nitrito 97.0-101.0%, pale yellow powder USP
Sodio nitrito

Sodium nitrite is used in many industrial processes, in meat curing, coloring, and preserving, and as a reagent in analytical chemistry.


It is used therapeutically as an antidote in cyanide poisoning. The compound is toxic and mutagenic and will react in vivo with secondary or tertiary amines thereby producing highly carcinogenic nitrosamines.


Soluble in water (820 mg/mL at 20 °C), methanol (4.5 mg/mL), ethanol (3 mg/mL), ether (slightly), and NH3 (very soluble)

Formula: NaNO₂
MW: 69 g/mol
Pto. Ebullición: 320
Pto. de fusión: 284
Número MDL: MFCD00011118
Núm. CAS: 7632-00-0
UN: 1500
ADR: 5.1 III
Índice Merck: 13,08721

Order Now

Resultados test especificaciones

Identity Test Passes
Loss on Drying (4 hours over silica) ≤0.25%
Heavy Metals ≤0,002%
Assay 97,0 - 101,0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR