EDTA disodium salt solution, 0,2% in phosphate buffered saline, MP Biomedicals
Proveedor: MP Biomedicals
092820549
ICNA092820549EA 24 EUR
ICNA092820549
EDTA disodium salt solution, 0,2% in phosphate buffered saline, MP Biomedicals
Tampones Tampones de clonación
EDTA disodium salt, (2%) solution, in phosphate buffered saline without calcium and magnesium. Used as a gentle non-enzymatic cell dissociation reagent.
  • Clear and colourless
  • pH: 4,0 to 6,0
  • Osmolality: 240 to 450 mM/kg