Chicken embryo extract, MP Biomedicals
Proveedor: MP Biomedicals
092850145
ICNA092850145EA 505 EUR
ICNA092850145
Chicken embryo extract, MP Biomedicals
Especímenes Especímenes preservados Preserved Animal Materials
Chicken embryo extract prepared from nine to 12 day old eggs.
  • Soluble in water
  • Brown solution with dirty aspect
  • Collected from registered flocks, which are free from clinical signs of notifiable diseases
Chicken embryo extract is prepared from 9 to 12 day old eggs collected from registered flocks and found to be free from clinical signs of notifiable diseases.
Información de suministro: Supplied in Dulbecco's PBS.