Microplate readers, multi-mode, SpectraMax® iD3
Proveedor: MOLECULAR DEVICES
ID3-STD ID3-INJ ID3INJSCACAD3 ID3INJSCACAD
735-0391EA 0 EUR
735-0391 735-0390 MLDVID3INJSCACAD3 ID3INJSCACAD
Microplate readers, multi-mode, SpectraMax® iD3
Lectores de microplacas
The SpectraMax® iD3 multi-mode microplate reader measures absorbance, fluorescence and luminescence and is ideal for busy multi-user, multi-application labs. Large, high resolution touchscreen interface allows you to easily set up methods, run experiments or view tutorial videos without the need for a dedicated computer workstation.
  • Easily and quickly pull up your custom protocols with a single tap, and reduce the possibility of confusion in busy, multi-user labs, with built-in near-field communication (NFC) functionality
  • Access your results quickly, with